Learn More
BioVendor, LLC Cc16 Anti-Hum Pab 0.1Mg BioVendor LLC
SDP
Supplier:  BioVendor, LLC RD18102222001
Encompass
TypePolyclonal AntibodyApplicationsWestern blotting, ELISA, Immunohistochemistry ource of AntigenHuman urineHostsRabbitPreparationThe antibody was raised in rabbits by immunization with the Human Club Cell Protein.Species ReactivityHumanPurification MethodImmunoaffinity chromatography on a column with immobilized recombinant Human Club Cell Protein (RD172022100, Biovendor).Antibody Content0.1 mg (determined by BCA method, BSA was used as a standard)FormulationThe antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.ReconstitutionAdd 0.2 ml of deionized water and let the lyophilized pellet dissolve completely. Slight turbidity may occur after reconstitution, which does not affect activity of the antibody. In this case clarify the solution by centrifugation.
Catalog No. 50-894-048