Learn More
Cayman Chemical ANTIBODY CCT020312 500ug
SDP
Catalog No. 502913900
Encompass_Preferred
An EIF2AK3 activator; increases phosphorylation of eIF2α at serine 51 in HT-29 and MCF-7 cells at 10 µM, an effect not seen in cells transfected with EIF2AK3 siRNA; reduces phosphorylation of Rb in HT-29 cells (EC50 = 5.1 µM); rapidly reduces the level of cyclin D1 in HT-29 cells at 10 µM; inhibits the growth of HT-29 colon carcinoma cells (GI50 = 2.1 µM)
Catalog No. 50-291-3900 Supplier Cayman Chemical Supplier No. 36577500 UG
An EIF2AK3 activator; increases phosphorylation of eIF2α at serine 51 in HT-29 and MCF-7 cells at 10 µM, an effect not seen in cells transfected with EIF2AK3 siRNA; reduces phosphorylation of Rb in HT-29 cells (EC50 = 5.1 µM); rapidly reduces the level of cyclin D1 in HT-29 cells at 10 µM; inhibits the growth of HT-29 colon carcinoma cells (GI50 = 2.1 µM)