Learn More
Cayman Chemical CD2665, Purity: ≥98%, CAS: 170355-78-9, 1 mg
SDP
Catalog No. 502971807
Encompass_Preferred
A selective RARβ and RARγ antagonist (Kds = 110, 306, and > 2,250 nM for RARγ, RARβ, and RARα, respectively, with no binding at RXRα)
Catalog No. 50-297-1807 Supplier Cayman Chemical Supplier No. 160311 MG
A selective RARβ and RARγ antagonist (Kds = 110, 306, and > 2,250 nM for RARγ, RARβ, and RARα, respectively, with no binding at RXRα)