Learn More
Cayman Chemical Ceapin-A7, Purity: ≥98%, CAS: 2323027-38-7, 25 mg
SDP
Catalog No. 502911771
Encompass_Preferred
An ATF6α inhibitor (IC50 = 0.59 µM in a reporter assay); selective for ATF6α over ATF6β at 18.9 µM; sensitizes U2OS cells to thapsigargin; chondrocyte-conditioned media from primary human chondrocytes pre-incubated with ceapin-A7 (0.5 µM) prior to stimulation with TNF-α, IFN-γ, or IL-17 has a reduced ability to induce tube formation in HUVECs; inhibits viral RNA production in Vero cells infected with SARS-CoV-2 at 15 µM
Catalog No. 50-291-1771 Supplier Cayman Chemical Supplier No. 3611325 MG
An ATF6α inhibitor (IC50 = 0.59 µM in a reporter assay); selective for ATF6α over ATF6β at 18.9 µM; sensitizes U2OS cells to thapsigargin; chondrocyte-conditioned media from primary human chondrocytes pre-incubated with ceapin-A7 (0.5 µM) prior to stimulation with TNF-α, IFN-γ, or IL-17 has a reduced ability to induce tube formation in HUVECs; inhibits viral RNA production in Vero cells infected with SARS-CoV-2 at 15 µM