Learn More
Cayman Chemical Cefaclor, Purity: ≥95%, CAS: 53994-73-3, 100 mg
SDP
Catalog No. 502964489
Encompass_Preferred
A cephalosporin antibiotic; active against S. pyogenes, S. pneumoniae, S. aureus, P. mirabilis, S. typhi, E. coli, and H. influenzae (MICs = 0.25, 0.25-2, 1-2, 1, 0.5, 1, and 2-4 μg/ml, respectively); protective against S. pyogenes, S. pneumoniae, S. aureus, P. mirabilis, S. typhi, E. coli, and H. influenzae infections in mice (ED50s = 0.08-30.2 mg/kg)
Catalog No. 50-296-4489 Supplier Cayman Chemical Supplier No. 23626100 MG
A cephalosporin antibiotic; active against S. pyogenes, S. pneumoniae, S. aureus, P. mirabilis, S. typhi, E. coli, and H. influenzae (MICs = 0.25, 0.25-2, 1-2, 1, 0.5, 1, and 2-4 μg/ml, respectively); protective against S. pyogenes, S. pneumoniae, S. aureus, P. mirabilis, S. typhi, E. coli, and H. influenzae infections in mice (ED50s = 0.08-30.2 mg/kg)