Learn More
Cayman Chemical Cefradine, Purity: ≥90%, CAS: 38821-53-3, 5 g
SDP
Catalog No. 502935814
Encompass_Preferred
An orally bioavailable β-lactam cephalosporin antibiotic; active against S. pyogenes, E. coli, K. pneumoniae, and E. cloacae (MICs = 0.04, 9.4, 9.4, and 6.3 μg/ml, respectively); active against clinical isolates of S. aureus (MICs = 0.8-6 μg/ml) and S. pyogenes (MICs = 0.1-0.4 μg/ml), as well as H. influenzae, E. coli, and K. pneumoniae (MICs = 6.2-12.5 μg/ml); increases survival in mouse models of systemic lethal infection by S. pyogenes, E. coli, K. pneumoniae, or E. cloacae (ED50s = 5, 37, 122, and 50 mg/kg, respectively), as well as by penicillin-susceptible or -resistant strains of S. aureus (ED50s = 18 and 91 mg/kg, respectively)
Catalog No. 50-293-5814 Supplier Cayman Chemical Supplier No. 260605 G
An orally bioavailable β-lactam cephalosporin antibiotic; active against S. pyogenes, E. coli, K. pneumoniae, and E. cloacae (MICs = 0.04, 9.4, 9.4, and 6.3 μg/ml, respectively); active against clinical isolates of S. aureus (MICs = 0.8-6 μg/ml) and S. pyogenes (MICs = 0.1-0.4 μg/ml), as well as H. influenzae, E. coli, and K. pneumoniae (MICs = 6.2-12.5 μg/ml); increases survival in mouse models of systemic lethal infection by S. pyogenes, E. coli, K. pneumoniae, or E. cloacae (ED50s = 5, 37, 122, and 50 mg/kg, respectively), as well as by penicillin-susceptible or -resistant strains of S. aureus (ED50s = 18 and 91 mg/kg, respectively)