Learn More
Cayman Chemical Celgosivir, Purity: ≥98%, CAS: 121104-96-9, 5 mg
SDP
Catalog No. 502912898
Encompass_Preferred
An α-glucosidase inhibitor (IC50 = 0.7 µg/ml) and a prodrug form of castanospermine; reduces BVDV levels in a plaque formation assay (IC50 = 16 mM) and increases survival of MDBK infected with BVDV (IC50 = 47 µM); reduces viremia and increases survival of mice infected with a lethal dose of dengue virus at 50 mg/kg twice daily, i.p.
Catalog No. 50-291-2898 Supplier Cayman Chemical Supplier No. 365265 MG
An α-glucosidase inhibitor (IC50 = 0.7 µg/ml) and a prodrug form of castanospermine; reduces BVDV levels in a plaque formation assay (IC50 = 16 mM) and increases survival of MDBK infected with BVDV (IC50 = 47 µM); reduces viremia and increases survival of mice infected with a lethal dose of dengue virus at 50 mg/kg twice daily, i.p.