Learn More
Cayman Chemical Centrinone, Purity: ≥95%, CAS: 1798871-30-3, 1 mg
SDP
Catalog No. 502936588
Encompass_Preferred
A Plk4 inhibitor (Ki = 0.16 nM); >1,100- and >2,800-fold selective for Plk4 over Aurora A and Aurora B kinases, respectively, and a panel of 442 kinases at 100 nM; depletes centrosomes and induces apoptosis in HeLa cells at 125 nM
Catalog No. 50-293-6588 Supplier Cayman Chemical Supplier No. 246841 MG
A Plk4 inhibitor (Ki = 0.16 nM); >1,100- and >2,800-fold selective for Plk4 over Aurora A and Aurora B kinases, respectively, and a panel of 442 kinases at 100 nM; depletes centrosomes and induces apoptosis in HeLa cells at 125 nM