Learn More
Cayman Chemical CGK733, Purity: ≥98%, CAS: 905973-89-9, 25 mg
SDP
Catalog No. 502969710
Encompass_Preferred
An anticancer compound; originally identified as an ATM and ATR kinase inhibitor in a study that has since been retracted; inhibits irradiation-induced ATM kinase activation in HEK293 cells at 10 µM but does not inhibit irradiation-induced phosphorylation of ATM or Chk2 in H460 or UV radiation-induced Chk1 phosphorylation at the same concentration; inhibits ATM kinase activity in a D54 glioblastoma mouse xenograft model at 25 mg/kg; reduces tumor growth in a CNE-2 nasopharyngeal carcinoma mouse xenograft model
Catalog No. 50-296-9710 Supplier Cayman Chemical Supplier No. 1624225 MG
An anticancer compound; originally identified as an ATM and ATR kinase inhibitor in a study that has since been retracted; inhibits irradiation-induced ATM kinase activation in HEK293 cells at 10 µM but does not inhibit irradiation-induced phosphorylation of ATM or Chk2 in H460 or UV radiation-induced Chk1 phosphorylation at the same concentration; inhibits ATM kinase activity in a D54 glioblastoma mouse xenograft model at 25 mg/kg; reduces tumor growth in a CNE-2 nasopharyngeal carcinoma mouse xenograft model