Learn More
Cayman Chemical Chlorin e6, Purity: ≥90%, CAS: 19660-77-6, 100 mg
SDP
Catalog No. 502964035
Encompass_Preferred
A photosensitizer; active against S. aureus, P. aeruginosa, E. coli, and S. typhimurium when used at 20 µM in combination with PDT; induces tumor regression when combined with PDT in several sarcoma rat xenograft models at 5 and 10 mg/kg
Catalog No. 50-296-4035 Supplier Cayman Chemical Supplier No. 21684100 MG
A photosensitizer; active against S. aureus, P. aeruginosa, E. coli, and S. typhimurium when used at 20 µM in combination with PDT; induces tumor regression when combined with PDT in several sarcoma rat xenograft models at 5 and 10 mg/kg