Learn More
Cayman Chemical Cilofexor, Purity: ≥98%, CAS: 1418274-28-8, 10 mg
SDP
Catalog No. 502936054
Encompass_Preferred
An FXR ligand; inhibits binding of a SRC-1-based synthetic peptide to FXR (EC50 = 50 = ≥100 nM)
Catalog No. 50-293-6054 Supplier Cayman Chemical Supplier No. 2574710 MG
An FXR ligand; inhibits binding of a SRC-1-based synthetic peptide to FXR (EC50 = 50 = ≥100 nM)