Learn More
Cayman Chemical Cisplatin, CAS: 15663-27-1, 100 mg
SDP
Catalog No. 502953147
Encompass_Preferred
A DNA-crosslinking agent; halts the cell cycle at the G2/M phase in vitro; active against murine tumors transplanted into mice and in mouse xenograft models; reduces tumor growth in a squamous cell carcinoma mouse xenograft model at doses of 7.5-12.5 mg/kg; inhibits RecA recombinase of M. tuberculosis (IC50 = 2 µM)
Catalog No. 50-295-3147 Supplier Cayman Chemical Supplier No. 13119100 MG
A DNA-crosslinking agent; halts the cell cycle at the G2/M phase in vitro; active against murine tumors transplanted into mice and in mouse xenograft models; reduces tumor growth in a squamous cell carcinoma mouse xenograft model at doses of 7.5-12.5 mg/kg; inhibits RecA recombinase of M. tuberculosis (IC50 = 2 µM)