Learn More
Cayman Chemical Citrulline, Purity: ≥95%, CAS: 372-75-8, 10 g
SDP
Catalog No. 502911380
Encompass_Preferred
A non-proteinogenic amino acid; produced by deimination of arginine through the post-translational modification citrullination
Catalog No. 50-291-1380 Supplier Cayman Chemical Supplier No. 3564810 G
A non-proteinogenic amino acid; produced by deimination of arginine through the post-translational modification citrullination