Learn More
Cayman Chemical CJ-13610, Purity: ≥98%, CAS: 179420-17-8, 10 mg
SDP
Catalog No. 502986371
Encompass_Preferred
An inhibitor of 5-LO (IC50 = 70 nM for 5-LO product formation in A23187-challenged PMNLs); inhibits recombinant 5-LO in a glutathione peroxidase-dependent manner (IC50 = 300 nM); reduces levels of LTB4 and decreases mechanical hyperalgesia in a rat model of chronic inflammatory pain induced by Freund's adjuvant (3-10 mg/kg); reverses tactile allodynia and increases hind paw weight bearing in a rat medial meniscal transection model of osteoarthritic pain (0.6-6 mg/kg)
Catalog No. 50-298-6371 Supplier Cayman Chemical Supplier No. 2176410 MG
An inhibitor of 5-LO (IC50 = 70 nM for 5-LO product formation in A23187-challenged PMNLs); inhibits recombinant 5-LO in a glutathione peroxidase-dependent manner (IC50 = 300 nM); reduces levels of LTB4 and decreases mechanical hyperalgesia in a rat model of chronic inflammatory pain induced by Freund's adjuvant (3-10 mg/kg); reverses tactile allodynia and increases hind paw weight bearing in a rat medial meniscal transection model of osteoarthritic pain (0.6-6 mg/kg)