Learn More
Cayman Chemical ANTIBODY CK-636 10mg
SDP
Catalog No. 502968996
Encompass_Preferred
A cell-permeable inhibitor of Arp2/3 complex action that inhibits actin polymerization in vitro, using human, bovine and fission yeast proteins (IC50s = 4, 32, and 24 µM, respectively); prevents the formation of actin filament comet tails by Listeria in infected SKOV3 cells (IC50 = 22 µM)
Catalog No. 50-296-8996 Supplier Cayman Chemical Supplier No. 1624310 MG
A cell-permeable inhibitor of Arp2/3 complex action that inhibits actin polymerization in vitro, using human, bovine and fission yeast proteins (IC50s = 4, 32, and 24 µM, respectively); prevents the formation of actin filament comet tails by Listeria in infected SKOV3 cells (IC50 = 22 µM)