Learn More
Cayman Chemical ANTIBODY CK-666 10mg
SDP
Catalog No. 502938023
Encompass_Preferred
An Arp2/3 inhibitor (IC50s = 4, 17, and 5 μM for the human, bovine, and S. pombe complexes, respectively, in an actin polymerization assay); binds at the Arp2/3 interface, stabilizing the inactive conformation of the complex; inhibits actin comet tail formation around intracellular L. monocytogenes in infected SKOV3 cells at 40 μM; induces actin remodeling, inhibits lamellipodia formation, and reduces the motility rate in M-1 epithelial cells at 200 μM; impairs long-term fear memory formation in rats when microinjected into the lateral amygdala at 100 μM
Catalog No. 50-293-8023 Supplier Cayman Chemical Supplier No. 2903810 MG
An Arp2/3 inhibitor (IC50s = 4, 17, and 5 μM for the human, bovine, and S. pombe complexes, respectively, in an actin polymerization assay); binds at the Arp2/3 interface, stabilizing the inactive conformation of the complex; inhibits actin comet tail formation around intracellular L. monocytogenes in infected SKOV3 cells at 40 μM; induces actin remodeling, inhibits lamellipodia formation, and reduces the motility rate in M-1 epithelial cells at 200 μM; impairs long-term fear memory formation in rats when microinjected into the lateral amygdala at 100 μM