Learn More
Cayman Chemical cKK-E12, Purity: ≥95%, CAS: 1432494-65-9, 25 mg
SDP
Catalog No. 502909542
Encompass_Preferred
An ionizable lipomer; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA; LNPs containing cKK-E12 and L-319 and encapsulating VZV mRNA encoding gE increase the lymph node and spleen levels of CD11c+CD80+ and CD11c+CD86+ cells and total serum IgG titers in mice, as well as decrease the number of VZV-induced rashes and the epithelial levels of VZV DNA in a guinea pig model of VZV-induced shingles
Catalog No. 50-290-9542 Supplier Cayman Chemical Supplier No. 3670025 MG
An ionizable lipomer; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA; LNPs containing cKK-E12 and L-319 and encapsulating VZV mRNA encoding gE increase the lymph node and spleen levels of CD11c+CD80+ and CD11c+CD86+ cells and total serum IgG titers in mice, as well as decrease the number of VZV-induced rashes and the epithelial levels of VZV DNA in a guinea pig model of VZV-induced shingles