Learn More
Cayman Chemical CL 387,785, Purity: ≥95%, CAS: 194423-06-8, 1 mg
SDP
Catalog No. 502983928
Encompass_Preferred
A potent, irreversible inhibitor of EGFR kinase activity (IC50 = 370 pM); halts cell cycling in cells that overexpress EGFR or c-ErbB2 (IC50s = 31-125 nM); profoundly blocks the growth of EGFR-overexpressing tumors in nude mice when given orally at 80 mg/kg/day for 10 days
Catalog No. 50-298-3928 Supplier Cayman Chemical Supplier No. 190811 MG
A potent, irreversible inhibitor of EGFR kinase activity (IC50 = 370 pM); halts cell cycling in cells that overexpress EGFR or c-ErbB2 (IC50s = 31-125 nM); profoundly blocks the growth of EGFR-overexpressing tumors in nude mice when given orally at 80 mg/kg/day for 10 days