Learn More
Cayman Chemical CM10, Purity: ≥98%, CAS: 692269-09-3, 5 mg
SDP
Catalog No. 502911954
Encompass_Preferred
A pan-ALDH1A inhibitor (IC50s = 1,700, 740, and 640 nM for human ALDH1A1, ALDH1A2, and ALDH1A3, respectively); selective for ALDH1A isoforms over ALDH2, -3A1, -4A1, -5A1, -1B1, and -1L1 at 20 µM; induces necroptosis in ovarian cancer stem cells
Catalog No. 50-291-1954 Supplier Cayman Chemical Supplier No. 361865 MG
A pan-ALDH1A inhibitor (IC50s = 1,700, 740, and 640 nM for human ALDH1A1, ALDH1A2, and ALDH1A3, respectively); selective for ALDH1A isoforms over ALDH2, -3A1, -4A1, -5A1, -1B1, and -1L1 at 20 µM; induces necroptosis in ovarian cancer stem cells