Learn More
Cayman Chemical CMP8, Purity: ≥98%, CAS: 851107-28-3, 1 mg
SDP
Catalog No. 502909701
Encompass_Preferred
A mutant ERα antagonist and ligand used for bump and hole; is selective for ERα containing three substitutions, ErαL384M, ErαM421G, and ErαG521R, over the wild-type ERα and ERβ (IC50s= 29, 1,085, and 2,232 nM, respectively); reduces reporter gene expression in HeLa cells expressing the triple mutant ERα (EC50 = 41 nM).
Catalog No. 50-290-9701 Supplier Cayman Chemical Supplier No. 365851 MG
A mutant ERα antagonist and ligand used for bump and hole; is selective for ERα containing three substitutions, ErαL384M, ErαM421G, and ErαG521R, over the wild-type ERα and ERβ (IC50s= 29, 1,085, and 2,232 nM, respectively); reduces reporter gene expression in HeLa cells expressing the triple mutant ERα (EC50 = 41 nM).