Learn More
Cayman Chemical Cobicistat, Purity: ≥98%, CAS: 1004316-88-4, 25 mg
SDP
Catalog No. 502964338
Encompass_Preferred
An inhibitor of CYP3A (IC50s = 30-285 nM for CYP3A metabolism of various HIV protease inhibitors); selective for CYP3A over other CYP isomers (IC50 = >25 μM for CYP1A2, 2C8, 2C9, and 2C19); does not inhibit HIV-1 protease (IC50 = >30 μM) or affect HIV replication in MT-2 cells (EC50 = >30 μM)
Catalog No. 50-296-4338 Supplier Cayman Chemical Supplier No. 2343325 MG
An inhibitor of CYP3A (IC50s = 30-285 nM for CYP3A metabolism of various HIV protease inhibitors); selective for CYP3A over other CYP isomers (IC50 = >25 μM for CYP1A2, 2C8, 2C9, and 2C19); does not inhibit HIV-1 protease (IC50 = >30 μM) or affect HIV replication in MT-2 cells (EC50 = >30 μM)