Learn More
BioVendor, LLC Cartilage Oligomeric Matrix Protein Human, Mouse Monoclonal Antibody, Clone: 12C4, 0.1 MG
SDP
Supplier:  BioVendor, LLC RD182080100C401
Encompass
TypeMonoclonal AntibodyApplicationsWestern blotting, ImmunohistochemistryAntibodies ApplicationsSource of AntigenHuman articular cartilageHostsMouseClone12C4PreparationThe antibody is a mouse monoclonal antibody against Human COMP.Purification MethodAffinity chromatography on a column with immobilized protein G.Antibody Content0.1 mg (determined by BCA method, BSA was used as a standard)FormulationThe antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.ReconstitutionAdd 0.2 ml of deionized water and let the lyophilized pellet dissolve completely. Slight turbidity may occur after reconstitution, which does not affect activity of the antibody. In this case clarify the solution by centrifugation.
Catalog No. 50-894-056