Learn More
Abcam Antibody Concentration And Clean-Up Kit, 3 Tests
Catalog No. NC0109770
Encompass_Preferred
The Antibody Concentration And Clean-Up Kit method allows the user to : -easily and quickly remove excess buffer from the antibody for a more concentrated antibody solution. -perform a simple buffer exchange to transfer the antibody into a more favorable buffer for conjugation
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0109770 Supplier Abcam Supplier No. AB1027783TEST
The Antibody Concentration And Clean-Up Kit method allows the user to : -easily and quickly remove excess buffer from the antibody for a more concentrated antibody solution. -perform a simple buffer exchange to transfer the antibody into a more favorable buffer for conjugation
The product is subject to the following: Abcam Restricted Use Statement