Learn More
Cayman Chemical Coumarin 6, Purity: ≥98%, CAS: 38215-36-0, 5 g
SDP
Catalog No. 502945927
Encompass_Preferred
An acid-sensitive fluorescent probe; upon protonation, it displays a shift in emission maxima from 410 nm to 510 nm upon excitation at 157 nm; has been commonly used in photolithography
Catalog No. 50-294-5927 Supplier Cayman Chemical Supplier No. 402145 G
An acid-sensitive fluorescent probe; upon protonation, it displays a shift in emission maxima from 410 nm to 510 nm upon excitation at 157 nm; has been commonly used in photolithography