Learn More
Cayman Chemical CpdA, Purity: ≥98%, CAS: 14593-25-0, 10 mg
SDP
Catalog No. 502995427
Encompass_Preferred
A non-steroidal selective glucocorticoid receptor modulator; contradictory results for binding affinity of CpdA compared with dexamethasone, either higher (4-fold, IC50 = 6.4 nM and 25.9 nM, respectively) or lower (63-fold, Kd = 81.8 nM and 1.29 nM, respectively) in whole cell binding assays
Catalog No. 50-299-5427 Supplier Cayman Chemical Supplier No. 2143810 MG
A non-steroidal selective glucocorticoid receptor modulator; contradictory results for binding affinity of CpdA compared with dexamethasone, either higher (4-fold, IC50 = 6.4 nM and 25.9 nM, respectively) or lower (63-fold, Kd = 81.8 nM and 1.29 nM, respectively) in whole cell binding assays