Learn More
Cayman Chemical CRT0066854, Purity: ≥98%, CAS: 1438881-19-6, 1 mg
SDP
Catalog No. 502939328
Encompass_Preferred
A PKCι and PKCζ inhibitor (IC50s = 132 and 639 nM, respectively); an inhibitor of ROCK2 (IC50 = 620 nM); is selective for PKCι, PKCζ, and ROCK2 over typical PKCs and 98 other kinases in a panel at 1 µM; decreases viability of A549 lung carcinoma cells (IC50 = 3.47 µM); decreases colony formation in HeLa cells by 65% at 1 µM; impairs lumen formation in MDCK cells in a Matrigel™ assay and migration of NRK-49F cells in a wound assay
Catalog No. 50-293-9328 Supplier Cayman Chemical Supplier No. 284631 MG
A PKCι and PKCζ inhibitor (IC50s = 132 and 639 nM, respectively); an inhibitor of ROCK2 (IC50 = 620 nM); is selective for PKCι, PKCζ, and ROCK2 over typical PKCs and 98 other kinases in a panel at 1 µM; decreases viability of A549 lung carcinoma cells (IC50 = 3.47 µM); decreases colony formation in HeLa cells by 65% at 1 µM; impairs lumen formation in MDCK cells in a Matrigel™ assay and migration of NRK-49F cells in a wound assay