Learn More
Cayman Chemical CRT0273750, Purity: ≥98%, CAS: 1979939-16-6, 5 mg
SDP
Catalog No. 502983901
Encompass_Preferred
An ENPP2 inhibitor (IC50 = 1 nM); prevents lysophosphatidylcholine hydrolysis in a plasma choline release assay (IC50 = 13 nM); inhibits the migration of 4T1 cells in migration assay (IC50 = 24 nM); decreases plasma levels of lysophosphatidic acid in an MDA-MB-231 mouse xenograft model
Catalog No. 50-298-3901 Supplier Cayman Chemical Supplier No. 209045 MG
An ENPP2 inhibitor (IC50 = 1 nM); prevents lysophosphatidylcholine hydrolysis in a plasma choline release assay (IC50 = 13 nM); inhibits the migration of 4T1 cells in migration assay (IC50 = 24 nM); decreases plasma levels of lysophosphatidic acid in an MDA-MB-231 mouse xenograft model