Learn More
Cayman Chemical CS 2100, Purity: ≥98%, CAS: 913827-99-3, 50 mg
SDP
Catalog No. 502970003
Encompass_Preferred
An S1P1 agonist; selectively induces GTPγS binding in CHO-K1 cells expressing human or rat S1P1 (EC50s = 4 and 1.5 nM, respectively) over those expressing human or rat S1P3 (EC50s = >20 and 7.4 µM, respectively); transiently reduces blood lymphocyte levels in rats with the levels returning to baseline within 24-48 hours at 0.1 and 1 mg/kg; reduces hind paw volume in a rat model of adjuvant-induced arthritis (ID50 = 0.44 mg/kg); decreases severity in a mouse model of EAE at 0.3 and 1 mg/kg
Catalog No. 50-297-0003 Supplier Cayman Chemical Supplier No. 1439650 MG
An S1P1 agonist; selectively induces GTPγS binding in CHO-K1 cells expressing human or rat S1P1 (EC50s = 4 and 1.5 nM, respectively) over those expressing human or rat S1P3 (EC50s = >20 and 7.4 µM, respectively); transiently reduces blood lymphocyte levels in rats with the levels returning to baseline within 24-48 hours at 0.1 and 1 mg/kg; reduces hind paw volume in a rat model of adjuvant-induced arthritis (ID50 = 0.44 mg/kg); decreases severity in a mouse model of EAE at 0.3 and 1 mg/kg