Learn More
Cayman Chemical CTP354, Purity: ≥99% (deutereated forms d1-d9), CAS: 1213669-91-0, 5 mg
SDP
Catalog No. 502914034
Encompass_Preferred
A deuterated derivative of L-838,417; binds to the GABAA receptor benzodiazepine binding site in non-α1 subunit-containing GABAA receptors; has a longer duration of action than L-838,417 in a rat model of neuropathic pain induced by sciatic nerve ligation
Catalog No. 50-291-4034 Supplier Cayman Chemical Supplier No. 366185 MG
A deuterated derivative of L-838,417; binds to the GABAA receptor benzodiazepine binding site in non-α1 subunit-containing GABAA receptors; has a longer duration of action than L-838,417 in a rat model of neuropathic pain induced by sciatic nerve ligation