Learn More
Cayman Chemical CU-115, Purity: ≥98%, CAS: 2471982-20-2, 5 mg
SDP
Catalog No. 502989306
Encompass_Preferred
A TLR8 antagonist (IC50 = 1.04 µM); selective for TLR8 over TLR7 (IC50 = >50 µM); inhibits increases in type I IFN transcriptional activity induced by 3p-hpRNA or G3-YSD in a luciferase reporter assay at 5 and 20 µM; decreases R-848-induced TNF-α and IL-1β production in THP-1 cells at 5 µM
Catalog No. 50-298-9306 Supplier Cayman Chemical Supplier No. 298155 MG
A TLR8 antagonist (IC50 = 1.04 µM); selective for TLR8 over TLR7 (IC50 = >50 µM); inhibits increases in type I IFN transcriptional activity induced by 3p-hpRNA or G3-YSD in a luciferase reporter assay at 5 and 20 µM; decreases R-848-induced TNF-α and IL-1β production in THP-1 cells at 5 µM