Learn More
Cayman Chemical CU-76, Purity: ≥95%, CAS: 2400954-58-5, 10 mg
SDP
Catalog No. 502990076
Encompass_Preferred
A cGAS inhibitor (IC50 = 0.24 µM); reduces DNA-induced dimerization of IRF3 in THP-1 cells; decreases IFN-stimulatory DNA-induced production of IFN-β in THP-1 cells at 10, 30, and 100 µM
Catalog No. 50-299-0076 Supplier Cayman Chemical Supplier No. 3103010 MG
A cGAS inhibitor (IC50 = 0.24 µM); reduces DNA-induced dimerization of IRF3 in THP-1 cells; decreases IFN-stimulatory DNA-induced production of IFN-β in THP-1 cells at 10, 30, and 100 µM