Learn More
Cayman Chemical CU-CPT8m, Purity: ≥98%, CAS: 125079-83-6, 10 mg
SDP
Catalog No. 502934212
Encompass_Preferred
An antagonist of TLR8 (Kd = 220 nM; IC50 = 67 nM in a reporter assay); is selective for TLR8 over a panel of all other human TLRs at 1 μM; inhibits increases in TNF-α and IL-8 mRNA expression induced by the TLR8 agonist R-848 in TLR8-overexpressing HEK-Blue cells at 1 μM; inhibits R-848-induced increases in TNF-α protein levels in differentiated THP-1 monocytes and human PBMCs in a concentration-dependent manner; decreases TNF-α and IL-8 levels in synovial cells derived from patients with osteoarthritis and TNF-α levels in PBMCs derived from patients with rheumatoid arthritis.
Catalog No. 50-293-4212 Supplier Cayman Chemical Supplier No. 2534910 MG
An antagonist of TLR8 (Kd = 220 nM; IC50 = 67 nM in a reporter assay); is selective for TLR8 over a panel of all other human TLRs at 1 μM; inhibits increases in TNF-α and IL-8 mRNA expression induced by the TLR8 agonist R-848 in TLR8-overexpressing HEK-Blue cells at 1 μM; inhibits R-848-induced increases in TNF-α protein levels in differentiated THP-1 monocytes and human PBMCs in a concentration-dependent manner; decreases TNF-α and IL-8 levels in synovial cells derived from patients with osteoarthritis and TNF-α levels in PBMCs derived from patients with rheumatoid arthritis.