Learn More
Cayman Chemical CVT-12012, Purity: ≥98%, CAS: 1018675-35-8, 25 mg
SDP
Catalog No. 502937668
Encompass_Preferred
An SCD inhibitor (IC50 = 6.1 nM in HepG2 cells); dose-dependently reduces levels of palmitoleic acid and vaccenic acid in the plasma and liver of rats fed a high-sucrose diet; reduces hepatic triglyceride levels in sucrose-fed rats at 20 mg/kg
Catalog No. 50-293-7668 Supplier Cayman Chemical Supplier No. 2715625 MG
An SCD inhibitor (IC50 = 6.1 nM in HepG2 cells); dose-dependently reduces levels of palmitoleic acid and vaccenic acid in the plasma and liver of rats fed a high-sucrose diet; reduces hepatic triglyceride levels in sucrose-fed rats at 20 mg/kg