Learn More
Cayman Chemical CVT-6883, Purity: ≥98%, CAS: 752222-83-6, 1 mg
SDP
Catalog No. 502947951
Encompass_Preferred
An adenosine A2B receptor antagonist (Ki = 22 nM); selective for adenosine A2B over adenosine A1, A2A, and A3 receptors (Kis = 1,940, 3,280, and 1,070 nM, respectively), as well as a panel of 84 proteins at 10 µM; decreases allergen challenge-induced BALF eosinophil and lymphocyte infiltration and airway reactivity in a mouse model of allergic asthma at 6 mg/kg; decreases fibrosis in the non-infarct zone, improves ejection fractions, and reduces ventricular tachycardia in a rat model of myocardial infarction
Catalog No. 50-294-7951 Supplier Cayman Chemical Supplier No. 333331 MG
An adenosine A2B receptor antagonist (Ki = 22 nM); selective for adenosine A2B over adenosine A1, A2A, and A3 receptors (Kis = 1,940, 3,280, and 1,070 nM, respectively), as well as a panel of 84 proteins at 10 µM; decreases allergen challenge-induced BALF eosinophil and lymphocyte infiltration and airway reactivity in a mouse model of allergic asthma at 6 mg/kg; decreases fibrosis in the non-infarct zone, improves ejection fractions, and reduces ventricular tachycardia in a rat model of myocardial infarction