Learn More
Cayman Chemical CW069, Purity: ≥95%, CAS: 1594094-64-0, 1 mg
SDP
Catalog No. 502964436
Encompass_Preferred
An inhibitor of KIFC/HSET (IC50 = 75 µM); increases the number of multipolar spindles in N1E-115 cells with supernumerary centrosomes at 100 and 200 µM; inhibits proliferation of N1E-115, but not NHDF, cells (IC50s = 86 and 181 µM, respectively)
Catalog No. 50-296-4436 Supplier Cayman Chemical Supplier No. 229361 MG
An inhibitor of KIFC/HSET (IC50 = 75 µM); increases the number of multipolar spindles in N1E-115 cells with supernumerary centrosomes at 100 and 200 µM; inhibits proliferation of N1E-115, but not NHDF, cells (IC50s = 86 and 181 µM, respectively)