Learn More
Cayman Chemical CX-4945, Purity: ≥98%, CAS: 1009820-21-6, 10 mg
SDP
Catalog No. 502968125
Encompass_Preferred
A potent, orally bioavailable inhibitor of casein kinase 2 (CK2; Ki = 0.38 nM); inhibits proliferation in a panel of cancer cell lines that overexpress CK2 and prevents tumor growth of breast and pancreatic cancer cell xenografts in mice
Catalog No. 50-296-8125 Supplier Cayman Chemical Supplier No. 1677910 MG
A potent, orally bioavailable inhibitor of casein kinase 2 (CK2; Ki = 0.38 nM); inhibits proliferation in a panel of cancer cell lines that overexpress CK2 and prevents tumor growth of breast and pancreatic cancer cell xenografts in mice