Learn More
Cayman Chemical ANTIBODY CXD101 500ug
SDP
Catalog No. 502983908
Encompass_Preferred
An HDAC1, HDAC2, and HDAC3 inhibitor (IC50s = 63, 570, and 550 nM, respectively); increases H3K9Ac in Colon-26 cells at 2.7 µM; decreases tumor volume, reduces tumor levels of macrophages, and increases tumor levels of Cd4+ and Cd8+ T cells in a Colon-26 murine colon cancer model at 50 mg/kg per day; increases serum levels of IgG and IgM antibodies thirty days after administration in mice immunized with the SARS-CoV-2 spike glycoprotein
Catalog No. 50-298-3908 Supplier Cayman Chemical Supplier No. 21159500 UG
An HDAC1, HDAC2, and HDAC3 inhibitor (IC50s = 63, 570, and 550 nM, respectively); increases H3K9Ac in Colon-26 cells at 2.7 µM; decreases tumor volume, reduces tumor levels of macrophages, and increases tumor levels of Cd4+ and Cd8+ T cells in a Colon-26 murine colon cancer model at 50 mg/kg per day; increases serum levels of IgG and IgM antibodies thirty days after administration in mice immunized with the SARS-CoV-2 spike glycoprotein