Learn More
Cayman Chemical CYM 5478, Purity: ≥98%, CAS: 870762-83-7, 1 mg
SDP
Catalog No. 502940144
Encompass_Preferred
An S1P2 agonist (EC50 = 0.78 μM in a reporter assay); selective for S1P2 over S1P1 and S1P3-5 in a TGFα-shedding assay (EC50s = 119, 1,690, 1,950, >10,000, and >10,000 nM, respectively); reduces serum starvation-induced decreases in C6 rat glioma cell viability from 0.1-10 μM; reduces cisplatin-induced accumulation of ROS and apoptosis in C6 cells at 10 μM
Catalog No. 50-294-0144 Supplier Cayman Chemical Supplier No. 290241 MG
An S1P2 agonist (EC50 = 0.78 μM in a reporter assay); selective for S1P2 over S1P1 and S1P3-5 in a TGFα-shedding assay (EC50s = 119, 1,690, 1,950, >10,000, and >10,000 nM, respectively); reduces serum starvation-induced decreases in C6 rat glioma cell viability from 0.1-10 μM; reduces cisplatin-induced accumulation of ROS and apoptosis in C6 cells at 10 μM