Learn More
Cayman Chemical CYM 5541, Purity: ≥98%, CAS: 945128-26-7, 50 mg
SDP
Catalog No. 502971760
Encompass_Preferred
A selective agonist of S1P3 (EC50 = 72-132 nM); less effectively activates S1P1 (EC50 = 28-38 µM) and is ineffective at S1P2, S1P4, and S1P5 as well as a panel of other receptors, ion channels and transporters
Catalog No. 50-297-1760 Supplier Cayman Chemical Supplier No. 1519050 MG
A selective agonist of S1P3 (EC50 = 72-132 nM); less effectively activates S1P1 (EC50 = 28-38 µM) and is ineffective at S1P2, S1P4, and S1P5 as well as a panel of other receptors, ion channels and transporters