Learn More
BioVendor, LLC Cystatin C Rat, Sheep Polyclonal Antibody, 0.1 MG
SDP
Supplier:  BioVendor, LLC RD384009100
Encompass_Preferred
TypePolyclonal AntibodyApplicationsWestern blotting, ELISA, ImmunohistochemistryAntibodies ApplicationsSource of AntigenE. coliHostsSheepIsotypeIgGPreparationThe antibody was raised in sheep by immunization with the recombinant Rat Cystatin C.Amino Acid SequenceRecombinant Rat Cystatin C, total 134 AA, Mw: 14.93 kDa (calculated). UniProtKB acc.no. P14841. N-terminal His-tag,14 extra AA .MRGSHHHHHHGMASGTSRPPPRLLGAPQEADASEEGVQRALDFAVSEYNKGSNDAYHSRAIQVVRARKQLVAGINYYLDVEMGRTTCTKSQTNLTNCPFHDQPHLMRKALCSFQIYSVPWKGTHTLTKSSCKNASpecies ReactivityRat. Not yet tested in other species.Purification MethodImmunoaffinity chromatography on a column with immobilized recombinant Rat Cystatin C.
Catalog No. 50-894-064