Learn More
Cayman Chemical CYT387, Purity: ≥95%, CAS: 1056634-68-4, 1 mg
SDP
Catalog No. 502972891
Encompass_Preferred
An ATP-competitive inhibitor of JAK1 and JAK2 (IC50s = 11 and 18 nM, respectively); causes growth suppression and apoptosis in JAK2-dependent hematopoietic cell lines; is efficacious in a mouse model of JAK2V617F-dependent myeloproliferative neoplasms
Catalog No. 50-297-2891 Supplier Cayman Chemical Supplier No. 162461 MG
An ATP-competitive inhibitor of JAK1 and JAK2 (IC50s = 11 and 18 nM, respectively); causes growth suppression and apoptosis in JAK2-dependent hematopoietic cell lines; is efficacious in a mouse model of JAK2V617F-dependent myeloproliferative neoplasms