Learn More
Cayman Chemical ANTIBODY CZC-8004 500ug
SDP
Catalog No. 502965408
Encompass_Preferred
An aminopyrimidine that binds a range of tyrosine kinases, including ABL, BTK, FAK, FER, JAK1, SRC, SYK, TEC, TNK1, TYK2, and YES
Catalog No. 50-296-5408 Supplier Cayman Chemical Supplier No. 16718500 UG
An aminopyrimidine that binds a range of tyrosine kinases, including ABL, BTK, FAK, FER, JAK1, SRC, SYK, TEC, TNK1, TYK2, and YES