Learn More
Cayman Chemical DS-3032, Purity: ≥98%, CAS: 1398568-47-2, 1 mg
SDP
Catalog No. 502943850
Encompass_Preferred
An inhibitor of the MDM2-p53 protein-protein interaction; induces apoptosis in WaGa and PeTa cells at 100 nM; decreases the viability of p53 wild-type MKL-1 cells but not p53 knockout MLK-1 cells at 1 µM; reduces tumor volume and increases survival in a PDX mouse model of Merkel cell carcinoma at 100 mg/kg per day
Catalog No. 50-294-3850 Supplier Cayman Chemical Supplier No. 418381 MG
An inhibitor of the MDM2-p53 protein-protein interaction; induces apoptosis in WaGa and PeTa cells at 100 nM; decreases the viability of p53 wild-type MKL-1 cells but not p53 knockout MLK-1 cells at 1 µM; reduces tumor volume and increases survival in a PDX mouse model of Merkel cell carcinoma at 100 mg/kg per day