Learn More
Cayman Chemical D-Ribulose, Purity: ≥95%, CAS: 488-84-6, 10 mg
SDP
Catalog No. 502980765
Encompass_Preferred
A ketopentose synthesized in the pentose phosphate pathway that plays a role in the formation of various bioactive compounds
Catalog No. 50-298-0765 Supplier Cayman Chemical Supplier No. 1889310 MG
A ketopentose synthesized in the pentose phosphate pathway that plays a role in the formation of various bioactive compounds