Learn More
Cayman Chemical D-Serine, Purity: ≥95%, CAS: 312-84-5, 250 g
SDP
Catalog No. 502991963
Encompass_Preferred
An NMDA co-agonist; potentiates glutamate-induced currents in oocytes expressing various subunit combinations (EC50s = 0.17-0.32 µM) more potently than glycine; reverses deficits in prepulse inhibition with prepulses of 69 and 73 dB, but not 81 dB, in mice with a mutant form of serine racemase (SrrY269*) at 600 mg/kg; increases the time spent in the target quadrant of the Morris water maze in mice with a mutant form of serine racemase (SrrY269*) at 600 mg/kg; serum levels are decreased in patients with schizophrenia
Catalog No. 50-299-1963 Supplier Cayman Chemical Supplier No. 31197250 G
An NMDA co-agonist; potentiates glutamate-induced currents in oocytes expressing various subunit combinations (EC50s = 0.17-0.32 µM) more potently than glycine; reverses deficits in prepulse inhibition with prepulses of 69 and 73 dB, but not 81 dB, in mice with a mutant form of serine racemase (SrrY269*) at 600 mg/kg; increases the time spent in the target quadrant of the Morris water maze in mice with a mutant form of serine racemase (SrrY269*) at 600 mg/kg; serum levels are decreased in patients with schizophrenia