Learn More
Cayman Chemical DS44960156, Purity: ≥98%, CAS: 2361327-08-2, 5 mg
SDP
Catalog No. 502940480
Encompass_Preferred
An MTHFD2 inhibitor (IC50 = 1.6 µM); selective for MTHFD2 over MTHFD1 (IC50 = >30 µM)
Catalog No. 50-294-0480 Supplier Cayman Chemical Supplier No. 283665 MG
An MTHFD2 inhibitor (IC50 = 1.6 µM); selective for MTHFD2 over MTHFD1 (IC50 = >30 µM)