Learn More
Cayman Chemical DADLE, Purity: ≥98%, CAS: 63631-40-3, 50 mg
SDP
Catalog No. 502940926
Encompass_Preferred
A peptide agonist of δ-opioid receptors (Ki = 2.06 nM in a radioligand binding assay); selective for δ-opioid over κ-opioid receptors (Ki = 16,000 nM) but also agonizes of μ-opioid receptors (Ki = 13.8 nM); inhibits electrically induced contractions in guinea pig myenteric plexus and mouse and rat vas deferens (IC50s = 8.9, 0.73, and 134 nM, respectively); induces analgesia in the tail-flick and hot plate tests in mice (ED50s = 0.03 and 0.027 nmol per animal, i.c.v.)
Catalog No. 50-294-0926 Supplier Cayman Chemical Supplier No. 2892850 MG
A peptide agonist of δ-opioid receptors (Ki = 2.06 nM in a radioligand binding assay); selective for δ-opioid over κ-opioid receptors (Ki = 16,000 nM) but also agonizes of μ-opioid receptors (Ki = 13.8 nM); inhibits electrically induced contractions in guinea pig myenteric plexus and mouse and rat vas deferens (IC50s = 8.9, 0.73, and 134 nM, respectively); induces analgesia in the tail-flick and hot plate tests in mice (ED50s = 0.03 and 0.027 nmol per animal, i.c.v.)