Learn More
Cayman Chemical Dapivirine, Purity: ≥98%, CAS: 244767-67-7, 25 mg
SDP
Catalog No. 502972890
Encompass_Preferred
A diarylpyrimidine derivative of the potent NNRTI efavirenz; inhibits HIV-1 reverse transcriptase (IC50= 24 nM: impedes the late stages of HIV-1 infection by interfering with HIV-1 Gag-Pol polyprotein processing
Catalog No. 50-297-2890 Supplier Cayman Chemical Supplier No. 1624725 MG
A diarylpyrimidine derivative of the potent NNRTI efavirenz; inhibits HIV-1 reverse transcriptase (IC50= 24 nM: impedes the late stages of HIV-1 infection by interfering with HIV-1 Gag-Pol polyprotein processing