Learn More
Cayman Chemical Darusentan, Purity: ≥98%, CAS: 171714-84-4, 1 mg
SDP
Catalog No. 502909208
Encompass_Preferred
An ETA receptor antagonist (Ki = 13 nM in rat aortic vascular smooth muscle cells); inhibits ET-1-induced contractions in isolated rat aortic rings and mesenteric microvessels (pA2s = 8.1 and 7.97, respectively); decreases systolic blood pressure in DOCA-salt hypertensive rats at 50 mg/kg; decreases MAP and LVDP in normotensive pigs at 1 and 5 mg/kg; reduces myocardial infarct size as a percentage of the area at risk in a porcine model of LAD coronary artery ligation-induced ischemia-reperfusion injury
Catalog No. 50-290-9208 Supplier Cayman Chemical Supplier No. 360341 MG
An ETA receptor antagonist (Ki = 13 nM in rat aortic vascular smooth muscle cells); inhibits ET-1-induced contractions in isolated rat aortic rings and mesenteric microvessels (pA2s = 8.1 and 7.97, respectively); decreases systolic blood pressure in DOCA-salt hypertensive rats at 50 mg/kg; decreases MAP and LVDP in normotensive pigs at 1 and 5 mg/kg; reduces myocardial infarct size as a percentage of the area at risk in a porcine model of LAD coronary artery ligation-induced ischemia-reperfusion injury