Learn More
Cayman Chemical Davercin, Purity: ≥98%, CAS: 55224-05-0, 10 mg
SDP
Catalog No. 502985029
Encompass_Preferred
A macrolide antibiotic; inhibits bacterial protein synthesis by targeting the 50S ribosomal subunit; has broad-spectrum activity against various Gram-positive and Gram-negative bacteria (MICs = 0.02-50 μg/ml); non-toxic to mice and rats (LD50s = 4.05 and 5.9 g/kg, respectively)
Catalog No. 50-298-5029 Supplier Cayman Chemical Supplier No. 2134810 MG
A macrolide antibiotic; inhibits bacterial protein synthesis by targeting the 50S ribosomal subunit; has broad-spectrum activity against various Gram-positive and Gram-negative bacteria (MICs = 0.02-50 μg/ml); non-toxic to mice and rats (LD50s = 4.05 and 5.9 g/kg, respectively)